Learn More
Cayman Chemical NG NG-dImethyl-L-ArgInIn 25mg
SDP
Catalog No. 502950514
Encompass_Preferred
An endogenous NOS inhibitor; levels are increased in Dahl salt-sensitive rats fed a high-salt diet concomittant with an increase in blood pressure
Catalog No. 50-295-0514 Supplier Cayman Chemical Supplier No. 3357625 MG
An endogenous NOS inhibitor; levels are increased in Dahl salt-sensitive rats fed a high-salt diet concomittant with an increase in blood pressure