Learn More
Cayman Chemical NG,NG-dimethyl-L-Arginine-d6 (hydrochloride), Purity: ≥99% deuterated forms (d1-d6), CAS: 1313730-20-9, 1 mg
SDP
Catalog No. 502950943
Encompass_Preferred
An internal standard for the quantification of NG,NG-dimethyl-L-arginine by GC- or LC-MS
Catalog No. 50-295-0943 Supplier Cayman Chemical Supplier No. 332661 MG
An internal standard for the quantification of NG,NG-dimethyl-L-arginine by GC- or LC-MS