Learn More
Medchemexpress LLC Ng-nitro-L-arginine | 2149-70-4 | 99.8% | 100 G
SDP
Catalog No. 5000231915
Encompass_Preferred
NG-nitro-L-arginine is a nitric oxide synthase (NOS) inhibitor used in biochemical and pharmacological research to modulate NO-dependent signaling and vascular responses. It is supplied in research-grade quantities for laboratory use.
  • Inhibits nitric oxide synthase for NO-related pathway studies.
  • Suitable for in vitro and in vivo research applications.
  • High purity (99.8%) for reproducible experimental results.
  • Molecular weight 219.20 g/mol; formula C6H13N5O4.
  • Available in bulk laboratory quantities for preparative or animal studies.
Catalog No. 50-002-31915 Supplier Medchemexpress LLC Supplier No. HY12115100G
NG-nitro-L-arginine is a nitric oxide synthase (NOS) inhibitor used in biochemical and pharmacological research to modulate NO-dependent signaling and vascular responses. It is supplied in research-grade quantities for laboratory use.
  • Inhibits nitric oxide synthase for NO-related pathway studies.
  • Suitable for in vitro and in vivo research applications.
  • High purity (99.8%) for reproducible experimental results.
  • Molecular weight 219.20 g/mol; formula C6H13N5O4.
  • Available in bulk laboratory quantities for preparative or animal studies.