Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NG Biotech NG-Test™ CTX-M
SDP

Catalog No. 23800539 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Form:
Liquid Ready-to-Use
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
23-800-539 Liquid Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-800-539 Supplier NG Biotech Supplier No. CTXM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

NG-Test™ CTX-M Multi, for rapid ESBL organism detection, FDA-cleared.

Rapid lateral flow test for detection of CTX-M enzymes (groups 1, 2, 8, 9, and 25) from pure colonies of Enterobacterales suspected of ESBL production.

Benefits:

  • Results in 15 minutes
  • Minimal hands-on time
  • Minimal training needs
  • No additional equipment needed
  • No maintenance costs
  • Stable at room temperature

5 Simple Steps:

  1. Add 5 drops of extraction buffer
  2. Collect sample
  3. Inoculate sample into buffer and mix until homogenous
  4. Draw sample into the pipette, until the liquid reaches the 100 μl line
  5. Dispense sample into the sample well

Specifications

Clia Complexity High
Detectable Analytes ESBL Organisms, CTX-M Enzymes from Group 1,2,8,9 and 25
Type LFA Assay
Disposable Single-use
For Use With (Application) Rapid ESBL Organism Detection
Form Liquid Ready-to-Use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.