Learn More
AdipoGen NG 011 (5 mg)
SDP
Supplier:  AdipoGen AGCN20178M005
Encompass_Preferred
Chemical. CAS: 141731-75-1. Formula: C32H38O15. MW: 662.7. Isolated from Penicillium sp. Antibiotic. Mycotoxin. Very weak cytotoxic activity. Antifungal. Inhibits cell wall biosynthesis. Nerve growth factor (NGF) potentiator.
Catalog No. 50-205-3568