Learn More
Electron Microscopy Sciences British Standard Graticle NG10, 21mm
SDP
Catalog No. 5031217
Encompass_Preferred
In this graticule the circle areas double progressively, hence the diameters alter by Root 2, so that the size classes can form a continuation of the standard series of sieves for particle sizing. Each particle is assigned to a size class defined by two adjacent circles, which represent the size limits of that class. Thus the distribution of size is obtained in terms of the diameter of circles having the same projected area as the particles. This method will cover particles in the range 150 micron to 0.38 micron. The size distributions with respect to their number and weight are determined separately. Final results are calculated as cumulative percents.
Originally designed by the National Coal Board for use in coal mining.
Size: 21 mm
Catalog No. 50-312-17 Supplier Electron Microscopy Sciences Supplier No. 6802621
In this graticule the circle areas double progressively, hence the diameters alter by Root 2, so that the size classes can form a continuation of the standard series of sieves for particle sizing. Each particle is assigned to a size class defined by two adjacent circles, which represent the size limits of that class. Thus the distribution of size is obtained in terms of the diameter of circles having the same projected area as the particles. This method will cover particles in the range 150 micron to 0.38 micron. The size distributions with respect to their number and weight are determined separately. Final results are calculated as cumulative percents.
Originally designed by the National Coal Board for use in coal mining.
Size: 21 mm