Learn More
Electron Microscopy Sciences New Porton Graticle NG12, 16mm
SDP
Catalog No. 5029273
Encompass_Preferred
The NG12 is particularly useful since the array of globes and circles are conveniently close to where the particles pass. At the end of each band of the sample the mechanical stage is traversed vertically to take in the next band until the whole sample has been covered.
Size: 16 mm
Catalog No. 50-292-73 Supplier Electron Microscopy Sciences Supplier No. 6803216
The NG12 is particularly useful since the array of globes and circles are conveniently close to where the particles pass. At the end of each band of the sample the mechanical stage is traversed vertically to take in the next band until the whole sample has been covered.
Size: 16 mm