Learn More
Alpco Ltd Canine NGAL ELISA
SDP
Supplier:  Alpco Ltd 54043
Encompass
The Canine NGAL ELISA is manufactured by BioPorto Diagnostics and intended for the determination of NGAL in dog urine, plasma, serum, tissue extracts, or culture media.
Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3628