Learn More
Crystal Chem Inc NGAL ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80729
Encompass
The NGAL ELISA is used to quantify levels of human NGAL in serum and plasma using only a 10 µL sample volume. Highlights: Kit uses sample size of only 10 µL. High sensitivity of 0.18 ng/mL. Dynamic range of 0.625 - 20 ng/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7848