Learn More
Enzo Life Sciences NGAL (human) ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences BPDKIT036
Encompass_Preferred
Highly senstitive ELISA, measuring as little as 4 pg/ml of NGAL. A colorimetric immunometric ELISA kit for the measurement of NGAL in human samples. Absorbance is read at 450nm. Alternative Name: Neutrophil gelatinase-associated lipocalin, Lipocalin 2, Siderocalin, Oncogene 24p3. Sensitivity: 4pg/ml (range 10-1000pg/ml). Assay Time: <4 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human. Contents: 1: 12 strips of 8 Microwells each coated with anti-NGAL (96 wells) + frame 2: Sample Diluent 3: NGAL Calibrators 1-8; 0, 10, 25, 50, 100, 250, 500, 1000 pg/mL 4: 25X Wash Solution Concentrate 5: Biotinylated NGAL Antibody 6: HRP-Streptavidin Conjugate 7: TMB Substrate 8: Stop Solution
Catalog No. 50-666-670