Learn More
Sino Biological NGAL / LCN2 / Lipocalin-2 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 80058MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat NGAL / LCN2 / Lipocalin-2 (rR NGAL / LCN2 / Lipocalin-2; Catalog#80058-R08B; P30152; Met1-Asn198). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8115