Learn More
Enzo Life Sciences NGAL (monkey) monoclonal antibody (56) (1 mg)
SDP
Supplier:  Enzo Life Sciences BPDABS063561
Encompass_Preferred
Clone: 56. Isotype: IgG1κ. Immunogen: Recombinant monkey NGAL. Species reactivity: Rhesus monkey, Cynomolgus. Applications: ELISA, IHC (PS). Purity: Protein A-affinity purified. Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.5M sodium chloride and 15mM sodium azide.
Catalog No. 50-201-2440