Learn More
Enzo Life Sciences NGAL (monkey) monoclonal antibody (68) (1 mg)
SDP
Supplier:  Enzo Life Sciences BPDABS063681
Encompass_Preferred
Clone: 68. Isotype: IgG1κ. Immunogen: Recombinant monkey NGAL. Species reactivity: Rhesus monkey, Cynomolgus. Applications: ELISA, IHC. Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.5M sodium chloride and 15mM sodium azide.
Catalog No. 50-201-2442