Learn More
Enzo Life Sciences NGAL (rat) ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences BPDKIT046
Encompass_Preferred
A colorimetric immunometric ELISA kit for the rapid measurement of NGAL in rat samples. Absorbance is read at 450nm. Alternative name: Neutrophil gelatinase-associated lipocalin, Lipocalin 2, Siderocalin, Oncogene 24p3. Assay Time: 4 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Rat .
Catalog No. 50-201-2476