Learn More
Enzo Life Sciences NGAL (rat) monoclonal antibody (32) (1 mg)
SDP
Supplier:  Enzo Life Sciences BPDABS039321
Encompass_Preferred
Clone: 32. Isotype: IgG1κ. Immunogen: Recombinant rat NGAL precursor. Species reactivity: Rat. Applications: ELISA, WB. Purity: Protein A/G-affinity purified. Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.5M sodium chloride and 15mM sodium azide.
Catalog No. 50-201-2416