Learn More
Enzo Life Sciences NGAL (rat), (recombinant) (1ml)
SDP
Catalog No. 502013909
Encompass_Preferred
Alternative name: Neutrophil gelatinase-associated lipocalin, Lipocalin 2, Siderocalin, Oncogene 24p3. Concentration: 400pg/ml. Formulation: Liquid. In 0.01 M phosphate buffer, pH 7.4, containing 0.14 M NaCl, 0.5% BSA, 0.5% Tween 20, 0.0016% Phenol red, and 0.1% Proclin 950. Source: Produced in E. coli. Long Term Storage: +4°C. Applications: ELISA .
Catalog No. 50-201-3909 Supplier Enzo Life Sciences Supplier No. ENZPRT1151000
Alternative name: Neutrophil gelatinase-associated lipocalin, Lipocalin 2, Siderocalin, Oncogene 24p3. Concentration: 400pg/ml. Formulation: Liquid. In 0.01 M phosphate buffer, pH 7.4, containing 0.14 M NaCl, 0.5% BSA, 0.5% Tween 20, 0.0016% Phenol red, and 0.1% Proclin 950. Source: Produced in E. coli. Long Term Storage: +4°C. Applications: ELISA .