Learn More
Cayman Chemical NG-amino-L-Arginine (hydrochloride), Purity: ≥97%, CAS: 1031799-40-2, 25 mg
SDP
Catalog No. 502888426
Encompass_Preferred
Inhibits nNOS, iNOS, and eNOS (Ki = 0.3, 3, and 2.5 μM, respectively); can be used both in cell culture and in vivo
Catalog No. 50-288-8426 Supplier Cayman Chemical Supplier No. 1055425 MG
Inhibits nNOS, iNOS, and eNOS (Ki = 0.3, 3, and 2.5 μM, respectively); can be used both in cell culture and in vivo