Learn More
Cayman Chemical Negamycin (hydrochloride), Purity: ≥80%, 5 mg
SDP
Catalog No. 502916820
Encompass_Preferred
An antibiotic; active against a variety of bacteria (3.12-100 µg/ml); stabilizes the interaction between tRNA and ribosomes and decreases the rate of mRNA translation; restores auditory brainstem response to sound tones in an mdx mouse model of DMD at 100 mg/kg
Catalog No. 50-291-6820 Supplier Cayman Chemical Supplier No. 377965 MG
An antibiotic; active against a variety of bacteria (3.12-100 µg/ml); stabilizes the interaction between tRNA and ribosomes and decreases the rate of mRNA translation; restores auditory brainstem response to sound tones in an mdx mouse model of DMD at 100 mg/kg