Learn More
Cayman Chemical NGB 2904 (hydrochloride), Purity: ≥98%, CAS: 189061-11-8, 10 mg
SDP
Catalog No. 502994036
Encompass_Preferred
A selective dopamine D3 receptor antagonist (Ki = 1.4 nM); attenuates cocaine- and stress-induced reinstatement of drug-seeking behavior in rats
Catalog No. 50-299-4036 Supplier Cayman Chemical Supplier No. 2145710 MG
A selective dopamine D3 receptor antagonist (Ki = 1.4 nM); attenuates cocaine- and stress-induced reinstatement of drug-seeking behavior in rats