Learn More
LAB BIOREAGENTS NGB III Buffer 250ml
Catalog No. 501959280
Encompass
Lab Bioreagents NGB (I, II, III) are newly developed next generation assay buffers that provides exceptional performance and broad compatibility for Western blot, ELISA, IHC and nucleic acid detection assays. The NGBs are basic buffers that hat requires neutralization by acid in the reaction. Among the tests, NGBs have been demonstrated to maintain the full bioactivity of proteins and antibodies.
Catalog No. 50-195-9280 Supplier LAB BIOREAGENTS Supplier No. NGB0030250
Lab Bioreagents NGB (I, II, III) are newly developed next generation assay buffers that provides exceptional performance and broad compatibility for Western blot, ELISA, IHC and nucleic acid detection assays. The NGBs are basic buffers that hat requires neutralization by acid in the reaction. Among the tests, NGBs have been demonstrated to maintain the full bioactivity of proteins and antibodies.