Learn More
TARGETMOL CHEMICALS INC NGD 94-1 5MG
SDP
Catalog No. 502877694
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. NGD 94-1 is a selective D4 receptor antagonist with an affinity of 3 nM for the D4 receptor and greater than 2 pM for the D1, D2, D3, and D5 receptors.NGD 94-1 can be used in the study of cognitive disorders and other psychiatric disorders. Purity 99.84%
Catalog No. 50-287-7694 Supplier TARGETMOL CHEMICALS INC Supplier No. T230665MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. NGD 94-1 is a selective D4 receptor antagonist with an affinity of 3 nM for the D4 receptor and greater than 2 pM for the D1, D2, D3, and D5 receptors.NGD 94-1 can be used in the study of cognitive disorders and other psychiatric disorders. Purity 99.84%