Learn More
ABclonal Technology NGDN Rabbit pAb
SDP
Supplier:  ABclonal Technology A14509500UL
Encompass_Preferred
Neuroguidin is an EIF4E (MIM 133440)-binding protein that interacts with CPEB (MIM 607342) and functions as a translational regulatory protein during development of the vertebrate nervous system (Jung et al., 2006 [PubMed 16705177]).
Catalog No. 50-001-06946