Learn More
Cayman Chemical ngelIs Salt 100mg
SDP
Catalog No. 502894636
Encompass_Preferred
A classical nitroxyl (NO-) donor; dissociates in a pH-dependent, first order process with a half-life of 2.3 minutes at 37°C, pH 7.4, to liberate 0.54 moles for NO per mole of parent compound
Catalog No. 50-289-4636 Supplier Cayman Chemical Supplier No. 82230100 MG
A classical nitroxyl (NO-) donor; dissociates in a pH-dependent, first order process with a half-life of 2.3 minutes at 37°C, pH 7.4, to liberate 0.54 moles for NO per mole of parent compound