Learn More
Cayman Chemical Angeli's Salt, Purity: ≥99%, CAS: 13826-64-7, 50 mg
SDP
Catalog No. 502896570
Encompass_Preferred
A classical nitroxyl (NO-) donor; dissociates in a pH-dependent, first order process with a half-life of 2.3 minutes at 37°C, pH 7.4, to liberate 0.54 moles for NO per mole of parent compound
Catalog No. 50-289-6570 Supplier Cayman Chemical Supplier No. 8223050 MG
A classical nitroxyl (NO-) donor; dissociates in a pH-dependent, first order process with a half-life of 2.3 minutes at 37°C, pH 7.4, to liberate 0.54 moles for NO per mole of parent compound