Learn More
Enzo Life Sciences Nerve growth factor 2.5S (mouse), (grade I) (10µg). CAS: 93828-24-6
SDP
Catalog No. 502009351
Encompass_Preferred
Alternative name: NGF 2.5S. Purity: ≥98% (SDS-PAGE). Formulation: Lyophilized. Source: Isolated from mouse submaxillary glands by a modification of the procedure described by T. Darling, et al.; Cell Culture Meth. Mol. Cell Biol. 4, 79 (1984). Long Term Storage: -20°C.
Catalog No. 50-200-9351 Supplier Enzo Life Sciences Supplier No. ALX521006C010
Alternative name: NGF 2.5S. Purity: ≥98% (SDS-PAGE). Formulation: Lyophilized. Source: Isolated from mouse submaxillary glands by a modification of the procedure described by T. Darling, et al.; Cell Culture Meth. Mol. Cell Biol. 4, 79 (1984). Long Term Storage: -20°C.