Learn More
Genscript Corporation Antibody Protein Proteins Proteins 1 MG 1/EA
Supplier:  Genscript Corporation Z031051
Encompass
NGF Receptor also known as Gp80-LNGFR p75 ICD CD271 and TNFRSF16 is a type I transmembrane protein belonging to the TNF receptor family. It is expressed by both neuronal and non-neuronal cells. Signaling through NGF Receptor has been shown to regulate gene expression cell migration and death. A truncated NGF Receptor containing only the extracellular domain has been detected in plasma amniotic fluid and urine and acts as a potent NGF antagonist.
Catalog No. 50-252-9835