Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4804MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: NGFR5 + NTR/912. Clonality: Monoclonal. Host: Mouse. Isotype: IgG. Positive Control: Neuronal axons, Schwann cells,, perineural cells of peripheral nerves, or tumors of nerve sheath differentiation e.g. Schwannoma, Neurofibroma. Soma, axons of sensory neurons,, ganglionic satellite cells. Melanomas. Cellular Staining: Cell surface, cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: NGFR from A875 melanoma cells (NGFR5); Recombinant human p75 NGFR protein (NTR/912). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 75kDa.Chromosome: 17q21-22. Gene Symbol: NGFR. Gene ID: 4804. Ensemble: ENSG00000064300. Uniprot: NP_002498.
Catalog No. 50-188-5017