Learn More
Aviva Systems Biology Corp NGFR Antibody - C-terminal region (ARP63279_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63279_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NGFR. It was validated on Western Blot. . Alternate names: CD271, Gp80-LNGFR, TNFRSF16, p75(NTR), p75NTRApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-929