Learn More
Medchemexpress LLC HY-117383 100mg , NGI-1 CAS:790702-57-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY117383100MG
Encompass_Preferred
HY-117383 100mg NGI-1 CAS: 790702-57-7 NGI-1 (ML414) is a potent oligosaccharyltransferase (OST) inhibitor, directly targeting and blocking the function of the OST catalytic subunits STT3A and STT3B. NGI-1 is a cell permeable inhibitor and can effectively reduce virus infectivity without affecting cell viability. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0577