Learn More
Medchemexpress LLC HY-117383 10mg Medchemexpress, NGI-1 CAS:790702-57-7 Purity:>98%
SDP
Catalog No. 501969469
Encompass_Preferred
Medchemexpress, HY-117383 10mg NGI-1 CAS:790702-57-7 NGI-1 (ML414) is a potent oligosaccharyltransferase (OST) inhibitor, directly targeting and blocking the function of the OST catalytic subunits STT3A and STT3B. NGI-1 is a cell permeable inhibitor and can effectively reduce virus infectivity without affecting cell viability. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9469 Supplier Medchemexpress LLC Supplier No. HY11738310MG
Medchemexpress, HY-117383 10mg NGI-1 CAS:790702-57-7 NGI-1 (ML414) is a potent oligosaccharyltransferase (OST) inhibitor, directly targeting and blocking the function of the OST catalytic subunits STT3A and STT3B. NGI-1 is a cell permeable inhibitor and can effectively reduce virus infectivity without affecting cell viability. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.