Learn More
Cayman Chemical ngIotensIn AtrIfluoroACTA 10mg
SDP
Catalog No. 502911917
Encompass_Preferred
An AT1 and AT2 receptor agonist and active metabolite of angiotensin II; binds to AT1 and AT2 receptors in HEK293 cells expressing the rat receptors (IC50s = 0.29 and 0.12 nM, respectively, in radioligand binding assays); induces calcium mobilization in VSMCs (EC50 = 100 nM); increases the perfusion pressure in isolated perfused rat kidney (EC50 = 443 nM), an effect that can be reversed by EXP-3174; the plasma Ang A/Ang II ratio is increased in patients with end-stage renal failure; serum levels are decreased in patients with pulmonary arterial hypertension
Catalog No. 50-291-1917 Supplier Cayman Chemical Supplier No. 3606310 MG
An AT1 and AT2 receptor agonist and active metabolite of angiotensin II; binds to AT1 and AT2 receptors in HEK293 cells expressing the rat receptors (IC50s = 0.29 and 0.12 nM, respectively, in radioligand binding assays); induces calcium mobilization in VSMCs (EC50 = 100 nM); increases the perfusion pressure in isolated perfused rat kidney (EC50 = 443 nM), an effect that can be reversed by EXP-3174; the plasma Ang A/Ang II ratio is increased in patients with end-stage renal failure; serum levels are decreased in patients with pulmonary arterial hypertension