Learn More
Cayman Chemical ngIotensIn Fragment 1-7ace 5mg
SDP
Catalog No. 502966093
Encompass_Preferred
An angiotensin metabolite that functions as a type 1 angiotensin II receptor agonist whereupon it can control hydroelectrolyte balance and demonstrates vasodilatory and anti-inflammatory actions in opposition to various adverse effects of angiotensin II
Catalog No. 50-296-6093 Supplier Cayman Chemical Supplier No. 175945 MG
An angiotensin metabolite that functions as a type 1 angiotensin II receptor agonist whereupon it can control hydroelectrolyte balance and demonstrates vasodilatory and anti-inflammatory actions in opposition to various adverse effects of angiotensin II