Learn More
Cayman Chemical ngIotensIn Fragment 17ace 10mg
SDP
Catalog No. 502966246
Encompass_Preferred
An angiotensin metabolite that functions as a type 1 angiotensin II receptor agonist whereupon it can control hydroelectrolyte balance and demonstrates vasodilatory and anti-inflammatory actions in opposition to various adverse effects of angiotensin II
Catalog No. 50-296-6246 Supplier Cayman Chemical Supplier No. 1759410 MG
An angiotensin metabolite that functions as a type 1 angiotensin II receptor agonist whereupon it can control hydroelectrolyte balance and demonstrates vasodilatory and anti-inflammatory actions in opposition to various adverse effects of angiotensin II