Learn More
Cayman Chemical ngIotensIn Ihumnratmo 10mg
SDP
Catalog No. 502935040
Encompass_Preferred
A peptide; is converted to the vasoconstrictor angiotensin II by ACE; binds to angiotensin receptors in human myometrium membrane preparations (Kd = 11.2 nM); increases the renal vascular resistance in isolated rat kidney (EC50 = 10.5 nM); increases MAP and plasma AVP level in fetal ewes (5 μg/kg, i.c.v.)
Catalog No. 50-293-5040 Supplier Cayman Chemical Supplier No. 2473710 MG
A peptide; is converted to the vasoconstrictor angiotensin II by ACE; binds to angiotensin receptors in human myometrium membrane preparations (Kd = 11.2 nM); increases the renal vascular resistance in isolated rat kidney (EC50 = 10.5 nM); increases MAP and plasma AVP level in fetal ewes (5 μg/kg, i.c.v.)