Learn More
Cayman Chemical ngIotensIn IIIhumnmouse 50mg
SDP
Catalog No. 502963967
Encompass_Preferred
An agonist at AT1 and AT2 receptors, with a 30-fold higher affinity for AT2
Catalog No. 50-296-3967 Supplier Cayman Chemical Supplier No. 2161850 MG
An agonist at AT1 and AT2 receptors, with a 30-fold higher affinity for AT2