Learn More
Cayman Chemical nglada oxbugna ngltn 10mg
SDP
Catalog No. 502951000
Encompass_Preferred
A flavonoid glycoside; scavenges ABTS radicals in a cell-free assay (IC50 = 18.13 UG/ml); induces apoptosis in MCF-7 and MDA-MB-231 human breast cancer cells at 2 UG/ml; inhibits LPS-induced production of IL-1β, IL-6, and NO in isolated rat chondrocytes, as well as reduces cartilage erosion and the loss of proteoglycan in the inflamed joints in a rat model of surgically induced osteoarthritis
Catalog No. 50-295-1000 Supplier Cayman Chemical Supplier No. 3465710 MG
A flavonoid glycoside; scavenges ABTS radicals in a cell-free assay (IC50 = 18.13 UG/ml); induces apoptosis in MCF-7 and MDA-MB-231 human breast cancer cells at 2 UG/ml; inhibits LPS-induced production of IL-1β, IL-6, and NO in isolated rat chondrocytes, as well as reduces cartilage erosion and the loss of proteoglycan in the inflamed joints in a rat model of surgically induced osteoarthritis