Learn More
Cayman Chemical Engeletin, Purity: ≥98%, CAS: 572-31-6, 50 mg
SDP
Catalog No. 502950029
Encompass_Preferred
A flavonoid glycoside; scavenges ABTS radicals in a cell-free assay (IC50 = 18.13 µg/ml); induces apoptosis in MCF-7 and MDA-MB-231 human breast cancer cells at 2 µg/ml; inhibits LPS-induced production of IL-1β, IL-6, and NO in isolated rat chondrocytes, as well as reduces cartilage erosion and the loss of proteoglycan in the inflamed joints in a rat model of surgically induced osteoarthritis
Catalog No. 50-295-0029 Supplier Cayman Chemical Supplier No. 3465750 MG
A flavonoid glycoside; scavenges ABTS radicals in a cell-free assay (IC50 = 18.13 µg/ml); induces apoptosis in MCF-7 and MDA-MB-231 human breast cancer cells at 2 µg/ml; inhibits LPS-induced production of IL-1β, IL-6, and NO in isolated rat chondrocytes, as well as reduces cartilage erosion and the loss of proteoglycan in the inflamed joints in a rat model of surgically induced osteoarthritis