Learn More
Cayman Chemical Marmesin, Purity: ≥98%, CAS: 13849-08-6, 10 mg
SDP
Catalog No. 502936210
Encompass_Preferred
A coumarin with anticancer activity; decreases proliferation and cell invasion induced by FBS in H1299 NSCLCs at 10 µM; decreases VEGF secretion in A549 and H1299 cells and inhibits capillary-like structure formation by HUVECs; decreases proliferation of U937 human leukemia cells (IC50 = 40 µM); reduces tumor growth in a U937 mouse xenograft model at 30 mg/kg
Catalog No. 50-293-6210 Supplier Cayman Chemical Supplier No. 2504910 MG
A coumarin with anticancer activity; decreases proliferation and cell invasion induced by FBS in H1299 NSCLCs at 10 µM; decreases VEGF secretion in A549 and H1299 cells and inhibits capillary-like structure formation by HUVECs; decreases proliferation of U937 human leukemia cells (IC50 = 40 µM); reduces tumor growth in a U937 mouse xenograft model at 30 mg/kg