Learn More
Cayman Chemical Suberosin, Purity: ≥98%, CAS: 581-31-7, 50 mg
SDP
Catalog No. 502910953
Encompass_Preferred
A prenylated coumarin with diverse biological activities; inhibits arachidonic acid-, collagen-, PAF-, ADP-, or U-46619-induced aggregation of washed isolated rabbit platelets (IC50s = 60, 132, 200, 234, and 200 µM, respectively); reduces arachidonic acid-induced TXB2 formation in the same model; inhibits PHA-induced NF-κB nuclear translocation in, as well as proliferation of, human PBMCs at 100 µM
Catalog No. 50-291-0953 Supplier Cayman Chemical Supplier No. 3503550 MG
A prenylated coumarin with diverse biological activities; inhibits arachidonic acid-, collagen-, PAF-, ADP-, or U-46619-induced aggregation of washed isolated rabbit platelets (IC50s = 60, 132, 200, 234, and 200 µM, respectively); reduces arachidonic acid-induced TXB2 formation in the same model; inhibits PHA-induced NF-κB nuclear translocation in, as well as proliferation of, human PBMCs at 100 µM