Learn More
Medchemexpress LLC NGM-313 1mg | 1mg
SDP
Catalog No. 5000324900
Encompass_Preferred
NGM-313 is a CHO-expressed human antibody targeting Klotho Beta NGM-313 contains a huIgG1 heavy chain and a hu light chain with a predicted molecular weight (MW) of 150 kDa The isotype control for NGM-313 can refer to Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-24900 Supplier Medchemexpress LLC Supplier No. HYP9906361MG
NGM-313 is a CHO-expressed human antibody targeting Klotho Beta NGM-313 contains a huIgG1 heavy chain and a hu light chain with a predicted molecular weight (MW) of 150 kDa The isotype control for NGM-313 can refer to Human IgG1 kappa Isotype Control (HY-P99001)