Learn More
Medchemexpress LLC NGM-621 1mg | 1mg
SDP
Catalog No. 5000324522
Encompass_Preferred
NGM-621 is a CHO-expressed human antibody that targets Complement C3 NGM-621 contains a huIgG1 heavy chain and a hu light chain with a predicted molecular weight (MW) of 149 04 kDa The isotype control for NGM-621 can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-24522 Supplier Medchemexpress LLC Supplier No. HYP9906371MG
NGM-621 is a CHO-expressed human antibody that targets Complement C3 NGM-621 contains a huIgG1 heavy chain and a hu light chain with a predicted molecular weight (MW) of 149 04 kDa The isotype control for NGM-621 can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)