Learn More
Electron Microscopy Sciences Mertz Graticle NGM1, 19mm
SDP
Catalog No. 5031224
Encompass_Preferred
Used to estimate the three-dimensional surface areas or the surface density of a component in a given volume, when the component does not have a random orientation. It comprises a test system with parallel curved lines used for measuring the intersection of points. Reference: W.A. Mertz. "Mikroskopic" Vol. 22 1967 pp 132-142.
Size: 19 mm
Catalog No. 50-312-24 Supplier Electron Microscopy Sciences Supplier No. 6803519
Used to estimate the three-dimensional surface areas or the surface density of a component in a given volume, when the component does not have a random orientation. It comprises a test system with parallel curved lines used for measuring the intersection of points. Reference: W.A. Mertz. "Mikroskopic" Vol. 22 1967 pp 132-142.
Size: 19 mm