Learn More
New England Biolabs, Inc. NgoMIV – 1000 units
SDP
Catalog No. 50812251
Encompass
A restriction endonuclease that recognizes the sequence G^CCGG_C.
Catalog No. 50-812-251 Supplier New England Biolabs, Inc. Supplier No. R0564S
A restriction endonuclease that recognizes the sequence G^CCGG_C.