Learn More
New England Biolabs, Inc. NgoMIV – 5000 units
SDP
Catalog No. 50812250
Encompass
A restriction endonuclease that recognizes the sequence G^CCGG_C.
Catalog No. 50-812-250 Supplier New England Biolabs, Inc. Supplier No. R0564L
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
A restriction endonuclease that recognizes the sequence G^CCGG_C.