Learn More
Aviva Systems Biology Corp NGRN Antibody - N-terminal region (ARP58748_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58748_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NGRN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NGRN. Alternate names: DSC92, NEUGRINApplicationWB. Species reactivity: Dog, Human, Rat
Catalog No. 50-014-62409