Learn More
Enzo Life Sciences AMPINEXT™ NGS Barcode (Index) Set-12 (144Reactions)
SDP
Catalog No. 502013793
Encompass_Preferred
The AMPINEXT™ NGS Barcode (Index) Set-12 is designed to construct multiplex DNA/RNA libraries used for next-generation sequencing with the Illumina platform including GAIIx, HiSeq, MiSeq, and NextSeq. This product includes 12 DNA barcodes, each of which contain an index and flow cell binding sequence and can be attached to the sample insert during library preparation. Multiplexing several samples in a single flow cell can be achieved by pooling with the EpiNext™ NGS Barcodes. Pooling barcoded samples into a single flow cell significantly reduces hands-on time and provides robust data quality in NGS.
Catalog No. 50-201-3793 Supplier Enzo Life Sciences Supplier No. ENZGEN5070144
The AMPINEXT™ NGS Barcode (Index) Set-12 is designed to construct multiplex DNA/RNA libraries used for next-generation sequencing with the Illumina platform including GAIIx, HiSeq, MiSeq, and NextSeq. This product includes 12 DNA barcodes, each of which contain an index and flow cell binding sequence and can be attached to the sample insert during library preparation. Multiplexing several samples in a single flow cell can be achieved by pooling with the EpiNext™ NGS Barcodes. Pooling barcoded samples into a single flow cell significantly reduces hands-on time and provides robust data quality in NGS.