Learn More
Meridian Life Science NGS Clean and Select Beads, 50ML
SDP
Catalog No. 5000856228
Encompass_Preferred
NGS Clean and Select Beads is an efficient NGS library preparation clean-up and selection system based on paramagnetic SPRI beads. It is designed to efficiently removing contaminants, such as nucleotides, primers, adapters, enzymes, buffer additives and salts, and for size selection of DNA fragments that can be used directly for downstream applications. The protocol can be used manually or adapted for liquid handling workstations.
Catalog No. 50-008-56228 Supplier Meridian Life Science Supplier No. MDX04150
NGS Clean and Select Beads is an efficient NGS library preparation clean-up and selection system based on paramagnetic SPRI beads. It is designed to efficiently removing contaminants, such as nucleotides, primers, adapters, enzymes, buffer additives and salts, and for size selection of DNA fragments that can be used directly for downstream applications. The protocol can be used manually or adapted for liquid handling workstations.