Learn More
LUNA NANOTECH INC Ngsprep Size Selection Magnetic Beads, 50 ML
Catalog No. 502566962
Encompass_Preferred
PuroMAG™ NGSPrep Magnetic Beads, 50 mL
The PuroMAG™ NGSPrep Magnetic Beads are designed for fast and highly effective DNA clean-up after polymerase chain reaction (PCR, qPCR, ddPCR), microarrays, sequencing and other types of ezymatic reactions by removing unincorporated primers, dNTPs (deoxynucleotide triphosphates), salts, and other impurities. Combination of SPRI technology and paramagnetic feature of the beads allows for specific size selection of DNA fragments during the clean-up procedure with high recovery of the DNA product. This is crucial application in library preparation for various sequencing techniques, including next-generation sequencing (NGS).
Features
- Easy and fast clean-up of DNA PCR samples with efficient recovery of the product
- Highly efficient size selection of purified fragmented genome or PCR product for NGS library prep
- High throughput clean-up of 96/384 samples
- Can be used o
Catalog No. 50-256-6962 Supplier LUNA NANOTECH INC Supplier No. NGSB50
PuroMAG™ NGSPrep Magnetic Beads, 50 mL
The PuroMAG™ NGSPrep Magnetic Beads are designed for fast and highly effective DNA clean-up after polymerase chain reaction (PCR, qPCR, ddPCR), microarrays, sequencing and other types of ezymatic reactions by removing unincorporated primers, dNTPs (deoxynucleotide triphosphates), salts, and other impurities. Combination of SPRI technology and paramagnetic feature of the beads allows for specific size selection of DNA fragments during the clean-up procedure with high recovery of the DNA product. This is crucial application in library preparation for various sequencing techniques, including next-generation sequencing (NGS).
Features
- Easy and fast clean-up of DNA PCR samples with efficient recovery of the product
- Highly efficient size selection of purified fragmented genome or PCR product for NGS library prep
- High throughput clean-up of 96/384 samples
- Can be used o