Learn More
Cayman Chemical nguInomycIn A 100ug
SDP
Catalog No. 502981389
Encompass_Preferred
An analog of leptomycin B that is highly cytotoxic to mouse P388 leukemia cells in vitro (IC50 = 0.1-0.2 ng/ml) as well as to Lewis lung carcinoma in mice
Catalog No. 50-298-1389 Supplier Cayman Chemical Supplier No. 20598100 UG
An analog of leptomycin B that is highly cytotoxic to mouse P388 leukemia cells in vitro (IC50 = 0.1-0.2 ng/ml) as well as to Lewis lung carcinoma in mice