Learn More
Electron Microscopy Sciences Weilbel 1 Graticle NGW1, 21mm
SDP
Catalog No. 5029704
Encompass_Preferred
Consists of 15 lines of equal length connecting the verticals of a regular hexagonal point network. Reference: E.R.Weibel Lab. Invest. Vol. 22 pp 131-152. "Principal and Methods for the Morphometric Study of the Lung and other Organs."
Size: 21 mm
Catalog No. 50-297-04 Supplier Electron Microscopy Sciences Supplier No. 6802221
Consists of 15 lines of equal length connecting the verticals of a regular hexagonal point network. Reference: E.R.Weibel Lab. Invest. Vol. 22 pp 131-152. "Principal and Methods for the Morphometric Study of the Lung and other Organs."
Size: 21 mm